Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACTGATCTAAAAGCAGAAAATTC[C/T]AGCTTTTGCTTTGTTTAGTGGATTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 166490 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SPP1 - secreted phosphoprotein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000582.2 | 258 | Intron | NP_000573.1 | |||
NM_001040058.1 | 258 | Intron | NP_001035147.1 | |||
NM_001040060.1 | 258 | Intron | NP_001035149.1 | |||
NM_001251829.1 | 258 | Intron | NP_001238758.1 | |||
NM_001251830.1 | 258 | Intron | NP_001238759.1 | |||
XM_017008564.1 | 258 | UTR 5 | XP_016864053.1 |