Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCTAGTGGCTACCATCATGGACAG[C/T]AATAGTCTATGGCATGAATAAACTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
STPG2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
STPG2 - sperm tail PG-rich repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174952.2 | Intron | NP_777612.1 | ||||
XM_011531886.2 | Intron | XP_011530188.1 | ||||
XM_017008049.1 | Intron | XP_016863538.1 | ||||
XM_017008050.1 | Intron | XP_016863539.1 | ||||
XM_017008051.1 | Intron | XP_016863540.1 |
STPG2-AS1 - STPG2 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |