Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCCAGGCGGCTGCTCGGCGTTCTC[C/T]CAGGTGACTGCTCGGAGTTCTCCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603028 | ||||||||||||||||||||
Literature Links: |
LOC100419170 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC100419170 - toll like receptor 2 pseudogene | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TLR2 - toll like receptor 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318787.1 | 86 | UTR 5 | NP_001305716.1 | |||
NM_001318789.1 | 86 | UTR 5 | NP_001305718.1 | |||
NM_001318790.1 | 86 | UTR 5 | NP_001305719.1 | |||
NM_001318791.1 | 86 | UTR 5 | NP_001305720.1 | |||
NM_001318793.1 | 86 | UTR 5 | NP_001305722.1 | |||
NM_001318795.1 | 86 | UTR 5 | NP_001305724.1 | |||
NM_001318796.1 | 86 | UTR 5 | NP_001305725.1 | |||
NM_003264.4 | 86 | UTR 5 | NP_003255.2 | |||
XM_011532215.2 | 86 | UTR 5 | XP_011530517.1 | |||
XM_011532216.2 | 86 | UTR 5 | XP_011530518.1 | |||
XM_017008573.1 | 86 | UTR 5 | XP_016864062.1 | |||
XM_017008574.1 | 86 | UTR 5 | XP_016864063.1 | |||
XM_017008575.1 | 86 | UTR 5 | XP_016864064.1 | |||
XM_017008576.1 | 86 | Intron | XP_016864065.1 |