Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACTTGAAAGACCCAGTGACCATCAA[C/T]TGTGGGCACAACTTCTGTCGCTCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
TRIM60 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TRIM60 - tripartite motif containing 60 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258025.1 | 254 | Silent Mutation | AAC,AAT | N,N 30 | NP_001244954.1 | |
NM_152620.2 | 254 | Silent Mutation | AAC,AAT | N,N 30 | NP_689833.1 | |
XM_011531683.2 | 254 | Silent Mutation | AAC,AAT | N,N 30 | XP_011529985.1 |