Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATGGACACCAGCCTAAGCTTTAG[A/G]CTGACACAAAAAGACTCCCTCCAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
VSTM4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VSTM4 - V-set and transmembrane domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031746.4 | 5288 | UTR 3 | NP_001026916.2 | |||
NM_144984.3 | 5288 | Intron | NP_659421.1 | |||
XM_017015825.1 | 5288 | Intron | XP_016871314.1 | |||
XM_017015826.1 | 5288 | Intron | XP_016871315.1 | |||
XM_017015827.1 | 5288 | Intron | XP_016871316.1 | |||
XM_017015828.1 | 5288 | UTR 3 | XP_016871317.1 |