Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGTGACAGTCATGTTCAAAGAAT[A/G]ATGTCCTACCAACTCCACAAAAAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
24 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605783 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BCAS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BCAS2 - breast carcinoma amplified sequence 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DENND2C - DENN domain containing 2C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001256404.1 | 3199 | Missense Mutation | CAT,TAT | H,Y 833 | NP_001243333.1 | |
NM_198459.3 | 3199 | Missense Mutation | CAT,TAT | H,Y 776 | NP_940861.3 |