Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGTTCTGTAATGATCAGTGATG[G/T]GATGAGGTAAGCAAATCTTAAGAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
C12orf42 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C12orf42 - chromosome 12 open reading frame 42 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099336.2 | Intron | NP_001092806.1 | ||||
NM_001278419.1 | Intron | NP_001265348.1 | ||||
NM_001278420.1 | Intron | NP_001265349.1 | ||||
NM_198521.3 | Intron | NP_940923.2 | ||||
XM_006719377.3 | Intron | XP_006719440.1 | ||||
XM_011538291.1 | Intron | XP_011536593.1 | ||||
XM_011538292.2 | Intron | XP_011536594.1 | ||||
XM_011538293.2 | Intron | XP_011536595.1 | ||||
XM_011538294.2 | Intron | XP_011536596.1 | ||||
XM_011538295.1 | Intron | XP_011536597.1 | ||||
XM_011538297.2 | Intron | XP_011536599.1 | ||||
XM_011538300.1 | Intron | XP_011536602.1 | ||||
XM_011538305.1 | Intron | XP_011536607.1 | ||||
XM_011538306.1 | Intron | XP_011536608.1 | ||||
XM_011538308.2 | Intron | XP_011536610.1 | ||||
XM_011538312.2 | Intron | XP_011536614.1 | ||||
XM_011538315.2 | Intron | XP_011536617.1 | ||||
XM_017019276.1 | Intron | XP_016874765.1 | ||||
XM_017019277.1 | Intron | XP_016874766.1 | ||||
XM_017019278.1 | Intron | XP_016874767.1 | ||||
XM_017019279.1 | Intron | XP_016874768.1 | ||||
XM_017019280.1 | Intron | XP_016874769.1 | ||||
XM_017019281.1 | Intron | XP_016874770.1 |