Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTCTAATTGTGGCTTGCTTACCATC[A/G]TGAGGCTGTATAACATAATGACTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 189904 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFYB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFYB - nuclear transcription factor Y subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006166.3 | 1016 | Silent Mutation | CAC,CAT | H,H 32 | NP_006157.1 | |
XM_005268908.4 | 1016 | Silent Mutation | CAC,CAT | H,H 33 | XP_005268965.1 | |
XM_005268909.4 | 1016 | Silent Mutation | CAC,CAT | H,H 33 | XP_005268966.1 | |
XM_005268910.4 | 1016 | Silent Mutation | CAC,CAT | H,H 33 | XP_005268967.1 |