Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAGGTCTCAGAATCCTCATTTCTTA[A/G]AAGCTCCCAGGTGACGTCTGTCAAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606231 MIM: 615748 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
APPL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
APPL2 - adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001251904.1 | Intron | NP_001238833.1 | ||||
NM_001251905.1 | Intron | NP_001238834.1 | ||||
NM_018171.3 | Intron | NP_060641.2 | ||||
XM_006719472.1 | Intron | XP_006719535.1 | ||||
XM_011538530.2 | Intron | XP_011536832.1 | ||||
XM_011538531.2 | Intron | XP_011536833.1 | ||||
XM_011538532.2 | Intron | XP_011536834.1 | ||||
XM_017019551.1 | Intron | XP_016875040.1 | ||||
XM_017019552.1 | Intron | XP_016875041.1 | ||||
XM_017019553.1 | Intron | XP_016875042.1 | ||||
XM_017019554.1 | Intron | XP_016875043.1 |
KIAA1033 - KIAA1033 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |