Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGCGGAAGCTGAACCACCAGAAC[A/G]TTGTCAAGCTCTTTGCGGTGGAGGA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 605048 MIM: 606524 | ||||||||||||||||||||
Literature Links: |
IKBKE PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
IKBKE - inhibitor of kappa light polypeptide gene enhancer in B-cells, kinase epsilon | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193321.1 | 777 | UTR 5 | NP_001180250.1 | |||
NM_001193322.1 | 777 | Missense Mutation | ATT,GTT | I,V 67 | NP_001180251.1 | |
NM_014002.3 | 777 | Missense Mutation | ATT,GTT | I,V 67 | NP_054721.1 | |
XM_005273356.2 | 777 | Missense Mutation | ATT,GTT | I,V 67 | XP_005273413.1 | |
XM_017002868.1 | 777 | UTR 5 | XP_016858357.1 |
MIR6769B - microRNA 6769b | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SRGAP2 - SLIT-ROBO Rho GTPase activating protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |