Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGAAAAGGAGAGGGTATGAAAAGG[C/T]AGACAGTTGAAGGGGTAGAGTCCAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
26 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS6KC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RPS6KC1 - ribosomal protein S6 kinase C1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001136138.2 | Intron | NP_001129610.1 | ||||
NM_001287218.1 | Intron | NP_001274147.1 | ||||
NM_001287219.1 | Intron | NP_001274148.1 | ||||
NM_001287220.1 | Intron | NP_001274149.1 | ||||
NM_001287221.1 | Intron | NP_001274150.1 | ||||
NM_012424.4 | Intron | NP_036556.2 | ||||
XM_005273095.1 | Intron | XP_005273152.1 | ||||
XM_005273096.1 | Intron | XP_005273153.1 | ||||
XM_006711272.1 | Intron | XP_006711335.1 | ||||
XM_017001024.1 | Intron | XP_016856513.1 | ||||
XM_017001025.1 | Intron | XP_016856514.1 | ||||
XM_017001026.1 | Intron | XP_016856515.1 | ||||
XM_017001027.1 | Intron | XP_016856516.1 | ||||
XM_017001028.1 | Intron | XP_016856517.1 | ||||
XM_017001029.1 | Intron | XP_016856518.1 | ||||
XM_017001030.1 | Intron | XP_016856519.1 | ||||
XM_017001031.1 | Intron | XP_016856520.1 | ||||
XM_017001032.1 | Intron | XP_016856521.1 | ||||
XM_017001033.1 | Intron | XP_016856522.1 | ||||
XM_017001034.1 | Intron | XP_016856523.1 |