Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTCACTTAATACAGCATTTAGAT[A/G]TTTAAAGTCATACACCTCATAAATA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
4 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 171835 MIM: 612023 | ||||||||||||||||||||
Literature Links: |
PFKFB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
PFKFB2 - 6-phosphofructo-2-kinase/fructose-2,6-biphosphatase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
YOD1 - YOD1 deubiquitinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276320.1 | 6210 | UTR 3 | NP_001263249.1 | |||
NM_018566.3 | 6210 | UTR 3 | NP_061036.3 |