Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTCACTGAGCACTTGTGGGGGCAC[A/G]ACCTCATCTTCTAAAGGCATCTTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602726 MIM: 108780 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLCN6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CLCN6 - chloride voltage-gated channel 6 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NPPA - natriuretic peptide A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006172.3 | 270 | Silent Mutation | GTC,GTT | V,V 57 | NP_006163.1 |
NPPA-AS1 - NPPA antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |