Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGGCTTTGCAGAGTTGAGTTGAG[A/G]TAATCAAGATGAACAAACAGTGCCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602000 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105373562 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105373562 - uncharacterized LOC105373562 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POLR1B - RNA polymerase I subunit B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001137604.2 | Intron | NP_001131076.1 | ||||
NM_001282772.1 | Intron | NP_001269701.1 | ||||
NM_001282774.1 | Intron | NP_001269703.1 | ||||
NM_001282776.1 | Intron | NP_001269705.1 | ||||
NM_001282777.1 | Intron | NP_001269706.1 | ||||
NM_001282779.1 | Intron | NP_001269708.1 | ||||
NM_019014.5 | Intron | NP_061887.2 |