Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603347 | ||||||||||||||||||||
Literature Links: |
NPAS2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
NPAS2 - neuronal PAS domain protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL31 - ribosomal protein L31 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000993.4 | 151 | Intron | NP_000984.1 | |||
NM_001098577.2 | 151 | Missense Mutation | ACC,GCC | T,A 22 | NP_001092047.1 | |
NM_001099693.1 | 151 | Intron | NP_001093163.1 |
TBC1D8 - TBC1 domain family member 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |