Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGAGGTCTACTTCCAAAGTCATAC[G/T]CTTAACCTGCCAGGCAACAATTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
VIT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
VIT - vitrin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001177969.1 | Intron | NP_001171440.1 | ||||
NM_001177970.1 | Intron | NP_001171441.1 | ||||
NM_001177971.1 | Intron | NP_001171442.1 | ||||
NM_001177972.1 | Intron | NP_001171443.1 | ||||
NM_053276.3 | Intron | NP_444506.2 | ||||
XM_011532905.1 | Intron | XP_011531207.1 | ||||
XM_011532906.1 | Intron | XP_011531208.1 | ||||
XM_017004325.1 | Intron | XP_016859814.1 | ||||
XM_017004326.1 | Intron | XP_016859815.1 | ||||
XM_017004327.1 | Intron | XP_016859816.1 | ||||
XM_017004328.1 | Intron | XP_016859817.1 |