Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGCCCCTCTCACCCCAGTAAAAAG[T/A]AGGAGAAAAAGAATACACAGATTAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
DHX57 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
DHX57 - DEAH-box helicase 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011533154.2 | 4609 | UTR 3 | XP_011531456.1 | |||
XM_011533155.2 | 4609 | UTR 3 | XP_011531457.1 | |||
XM_011533156.2 | 4609 | Intron | XP_011531458.1 | |||
XM_011533157.1 | 4609 | Intron | XP_011531459.1 |