Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGGTCTGCAGAGACAGAATTAAA[A/C]AGCTGCATAATCAAGAGAGTGACAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605771 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
COX7A2L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
COX7A2L - cytochrome c oxidase subunit 7A2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001319036.1 | Intron | NP_001305965.1 | ||||
NM_001319037.1 | Intron | NP_001305966.1 | ||||
NM_001319038.1 | Intron | NP_001305967.1 | ||||
NM_001319040.1 | Intron | NP_001305969.1 | ||||
NM_004718.3 | Intron | NP_004709.2 |