Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTGGGCTGAAAAATTTGCATAATA[A/C]ATACTGTATTTGTAATGTAAATTAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613580 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DBIL5P2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DBIL5P2 - diazepam binding inhibitor-like 5 pseudogene 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDPCP - WD repeat containing planar cell polarity effector | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042692.2 | 3145 | Intron | NP_001036157.1 | |||
NM_015910.5 | 3145 | Intron | NP_056994.3 | |||
XM_005264348.3 | 3145 | UTR 3 | XP_005264405.1 | |||
XM_011532881.2 | 3145 | UTR 3 | XP_011531183.1 | |||
XM_011532884.2 | 3145 | Intron | XP_011531186.1 | |||
XM_011532886.2 | 3145 | Intron | XP_011531188.1 | |||
XM_011532887.2 | 3145 | Intron | XP_011531189.1 | |||
XM_011532890.2 | 3145 | Intron | XP_011531192.1 | |||
XM_011532891.2 | 3145 | Intron | XP_011531193.1 | |||
XM_017004251.1 | 3145 | UTR 3 | XP_016859740.1 | |||
XM_017004252.1 | 3145 | Intron | XP_016859741.1 | |||
XM_017004253.1 | 3145 | Intron | XP_016859742.1 | |||
XM_017004254.1 | 3145 | Intron | XP_016859743.1 |