Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGAAAAGGGACAGGACCAGAAAAGT[A/G]ACTATTGGGTACTAGGCTTAGTACC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610052 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CHMP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CHMP3 - charged multivesicular body protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001005753.2 | 2592 | UTR 3 | NP_001005753.1 | |||
NM_001193517.1 | 2592 | UTR 3 | NP_001180446.1 | |||
NM_016079.3 | 2592 | UTR 3 | NP_057163.1 |
RNF103-CHMP3 - RNF103-CHMP3 readthrough | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198954.1 | 2592 | UTR 3 | NP_001185883.1 |