Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATTTGGTTCCACAAGGGGGACAC[C/T]TTGGGGTTCACAAGGATGGGGGCCA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 186910 MIM: 186730 | ||||||||||||||||||||
Literature Links: |
CD8A PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CD8A - CD8a molecule | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
CD8B - CD8b molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178100.1 | 843 | UTR 3 | NP_001171571.1 | |||
NM_004931.4 | 843 | Intron | NP_004922.1 | |||
NM_172101.3 | 843 | UTR 3 | NP_742099.1 | |||
NM_172102.3 | 843 | UTR 3 | NP_742100.1 | |||
NM_172213.3 | 843 | UTR 3 | NP_757362.1 | |||
XM_011533164.2 | 843 | Intron | XP_011531466.1 |