Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGTAAAGGATGAGGAATGTAGC[C/T]CGTCATTTACAACGTGCATTCACTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611479 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CCDC121 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CCDC121 - coiled-coil domain containing 121 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142683.2 | Intron | NP_001136155.1 | ||||
NM_024584.4 | Intron | NP_078860.2 | ||||
XM_005264560.3 | Intron | XP_005264617.1 |
GPN1 - GPN-loop GTPase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145047.1 | Intron | NP_001138519.1 | ||||
NM_001145048.1 | Intron | NP_001138520.1 | ||||
NM_001145049.1 | Intron | NP_001138521.1 | ||||
NM_007266.3 | Intron | NP_009197.2 |
ZNF512 - zinc finger protein 512 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |