Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTCCGTTAATTTTAGAATGAGAGC[A/G]CTCGCTGTGCTGTCTGTCACGCTGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606765 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPO PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
TPO - thyroid peroxidase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000547.5 | 95 | Silent Mutation | GCA,GCG | A,A 3 | NP_000538.3 | |
NM_001206744.1 | 95 | Silent Mutation | GCA,GCG | A,A 3 | NP_001193673.1 | |
NM_001206745.1 | 95 | Silent Mutation | GCA,GCG | A,A 3 | NP_001193674.1 | |
NM_175719.3 | 95 | Silent Mutation | GCA,GCG | A,A 3 | NP_783650.1 | |
NM_175721.3 | 95 | Silent Mutation | GCA,GCG | A,A 3 | NP_783652.1 | |
NM_175722.3 | 95 | Silent Mutation | GCA,GCG | A,A 3 | NP_783653.1 | |
XM_011510379.2 | 95 | Silent Mutation | GCA,GCG | A,A 3 | XP_011508681.1 | |
XM_011510380.2 | 95 | Silent Mutation | GCA,GCG | A,A 3 | XP_011508682.1 | |
XM_011510381.2 | 95 | Silent Mutation | GCA,GCG | A,A 3 | XP_011508683.1 |