Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAACAAGCAAGTACTGGGCTTCCG[C/T]GCGCTCTGGACTGCCTGAAGTTAAT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 608868 MIM: 611037 | |||||||||||||||||||||||
Literature Links: |
LRIG1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LRIG1 - leucine rich repeats and immunoglobulin like domains 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_015541.2 | 3688 | Missense Mutation | ACG,GCG | T,A 1050 | NP_056356.2 | |
XM_011533578.1 | 3688 | Missense Mutation | ACG,GCG | T,A 1002 | XP_011531880.1 | |
XM_011533579.2 | 3688 | Missense Mutation | ACG,GCG | T,A 790 | XP_011531881.1 | |
XM_017006134.1 | 3688 | Missense Mutation | ACG,GCG | T,A 1025 | XP_016861623.1 | |
XM_017006135.1 | 3688 | Missense Mutation | ACG,GCG | T,A 824 | XP_016861624.1 | |
XM_017006136.1 | 3688 | Missense Mutation | ACG,GCG | T,A 790 | XP_016861625.1 |
SLC25A26 - solute carrier family 25 member 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164796.1 | 3688 | Intron | NP_001158268.1 | |||
NM_173471.3 | 3688 | Intron | NP_775742.4 | |||
XM_006712956.1 | 3688 | Intron | XP_006713019.1 | |||
XM_011533327.2 | 3688 | Intron | XP_011531629.1 | |||
XM_011533328.1 | 3688 | Intron | XP_011531630.1 | |||
XM_017005671.1 | 3688 | Intron | XP_016861160.1 | |||
XM_017005672.1 | 3688 | Intron | XP_016861161.1 | |||
XM_017005673.1 | 3688 | Intron | XP_016861162.1 | |||
XM_017005674.1 | 3688 | Intron | XP_016861163.1 |