Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTACGAGGTACACAGAGGGCTATA[A/G]TCAGAGATCGAGCAGCTTTAGAGAA
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 609512 | ||||||||||||||||||||
Literature Links: |
CHMP2B PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CHMP2B - charged multivesicular body protein 2B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001244644.1 | 176 | Intron | NP_001231573.1 | |||
NM_014043.3 | 176 | Missense Mutation | ATC,GTC | I,V 29 | NP_054762.2 | |
XM_011533576.2 | 176 | Missense Mutation | ATC,GTC | I,V 45 | XP_011531878.1 |