Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAAGTAATGTTCAGTATAATAAAA[C/T]CCAAATATAAACCAATTAAAAACTT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 606568 | |||||||||||||||||||||||
Literature Links: |
LZTFL1 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
LZTFL1 - leucine zipper transcription factor like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001276378.1 | Intron | NP_001263307.1 | ||||
NM_001276379.1 | Intron | NP_001263308.1 | ||||
NM_020347.3 | Intron | NP_065080.1 | ||||
XM_006713207.2 | Intron | XP_006713270.2 | ||||
XM_011533838.2 | Intron | XP_011532140.1 | ||||
XM_017006644.1 | Intron | XP_016862133.1 | ||||
XM_017006645.1 | Intron | XP_016862134.1 |