Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTCATACAACCAGCCGGGCCAACA[C/T]ATTGAGCCTTAGAATTAGGGCAGCT
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 612402 MIM: 607950 MIM: 607237 | |||||||||||||||||||||||
Literature Links: |
ALS2CL PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
ALS2CL - ALS2 C-terminal like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PRSS50 - protease, serine 50 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TMIE - transmembrane inner ear | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_147196.2 | Intron | NP_671729.2 | ||||
XM_006713097.3 | Intron | XP_006713160.1 |