Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATAAAGGATGTCTCAAAAGGCTTAG[C/T]GCAGTTTTAGCTTTAATAACTTCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRP2BP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRP2BP - LRP2 binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018409.3 | 1709 | UTR 3 | NP_060879.2 | |||
XM_005263125.4 | 1709 | UTR 3 | XP_005263182.1 | |||
XM_006714260.3 | 1709 | UTR 3 | XP_006714323.1 | |||
XM_006714265.3 | 1709 | UTR 3 | XP_006714328.1 | |||
XM_011532110.2 | 1709 | UTR 3 | XP_011530412.1 | |||
XM_011532111.2 | 1709 | UTR 3 | XP_011530413.1 | |||
XM_017008410.1 | 1709 | UTR 3 | XP_016863899.1 | |||
XM_017008411.1 | 1709 | UTR 3 | XP_016863900.1 |
SNX25 - sorting nexin 25 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001317781.1 | 1709 | Intron | NP_001304710.1 |