Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAAGTAACTGTTCCATCGAGGCCTG[C/T]AGCTCTTGTAAATCTGTGTCAGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604588 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NEK1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NEK1 - NIMA related kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199397.1 | 3907 | Silent Mutation | NP_001186326.1 | |||
NM_001199398.1 | 3907 | Silent Mutation | NP_001186327.1 | |||
NM_001199399.1 | 3907 | Silent Mutation | NP_001186328.1 | |||
NM_001199400.1 | 3907 | Silent Mutation | NP_001186329.1 | |||
NM_012224.2 | 3907 | Silent Mutation | NP_036356.1 | |||
XM_006714228.1 | 3907 | Silent Mutation | XP_006714291.1 | |||
XM_011532003.1 | 3907 | Silent Mutation | XP_011530305.1 | |||
XM_011532004.1 | 3907 | Silent Mutation | XP_011530306.1 | |||
XM_011532005.2 | 3907 | Intron | XP_011530307.1 | |||
XM_017008249.1 | 3907 | Silent Mutation | XP_016863738.1 | |||
XM_017008250.1 | 3907 | Silent Mutation | XP_016863739.1 | |||
XM_017008251.1 | 3907 | Silent Mutation | XP_016863740.1 | |||
XM_017008252.1 | 3907 | Silent Mutation | XP_016863741.1 | |||
XM_017008253.1 | 3907 | Silent Mutation | XP_016863742.1 | |||
XM_017008254.1 | 3907 | Silent Mutation | XP_016863743.1 |