Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATCCATGGAAAGAAATGTTGGTA[C/T]CCTGAACTTAAAATGAAAAATAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611754 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AADAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AADAT - aminoadipate aminotransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286682.1 | 1069 | Missense Mutation | ATA,GTA | I,V 247 | NP_001273611.1 | |
NM_001286683.1 | 1069 | Missense Mutation | ATA,GTA | I,V 243 | NP_001273612.1 | |
NM_016228.3 | 1069 | Missense Mutation | ATA,GTA | I,V 243 | NP_057312.1 | |
NM_182662.1 | 1069 | Missense Mutation | ATA,GTA | I,V 243 | NP_872603.1 | |
XM_006714231.2 | 1069 | Missense Mutation | ATA,GTA | I,V 282 | XP_006714294.1 | |
XM_011532020.2 | 1069 | Missense Mutation | ATA,GTA | I,V 128 | XP_011530322.1 |