Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGAATGACAAAAAGCTGGGTTGAG[A/G]AAGAGGCTCCCTGCTCTGACTGTTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603640 MIM: 609383 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ADAM19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ADAM19 - ADAM metallopeptidase domain 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_033274.4 | 3356 | UTR 3 | NP_150377.1 | |||
XM_005266003.2 | 3356 | Intron | XP_005266060.1 | |||
XM_011534682.2 | 3356 | UTR 3 | XP_011532984.1 | |||
XM_017010009.1 | 3356 | UTR 3 | XP_016865498.1 |
NIPAL4 - NIPA like domain containing 4 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |