Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGTGTACAAGGTGTCCCTGTGGCCC[A/G]CTGCCTCAGCTTTAGCCACCTTTAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607352 MIM: 603612 | ||||||||||||||||||||
Literature Links: |
RHOBTB2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RHOBTB2 - Rho related BTB domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TNFRSF10B - TNF receptor superfamily member 10b | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003842.4 | 1333 | Missense Mutation | GCG,GTG | A,V 376 | NP_003833.4 | |
NM_147187.2 | 1333 | Missense Mutation | GCG,GTG | A,V 347 | NP_671716.2 | |
XM_017013944.1 | 1333 | Missense Mutation | GCG,GTG | A,V 311 | XP_016869433.1 |