Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCTCTCCCGCTGTGTTTTGCTGTA[A/G]ATCTTCTCCACTGGCACCCTGAACC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 606387 | ||||||||||||||||||||
Literature Links: |
TOP1MT PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
TOP1MT - topoisomerase (DNA) I, mitochondrial | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001258446.1 | 1767 | Silent Mutation | NP_001245375.1 | |||
NM_001258447.1 | 1767 | Silent Mutation | NP_001245376.1 | |||
NM_052963.2 | 1767 | Silent Mutation | NP_443195.1 | |||
XM_005250781.3 | 1767 | Intron | XP_005250838.1 |
ZNF696 - zinc finger protein 696 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |