Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGTGTTGCTTCCGTGCTGAGATCAC[A/G]TTCTTTGTTTTGTTTTGTGCCCGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608136 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARHGEF10 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARHGEF10 - Rho guanine nucleotide exchange factor 10 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001308152.1 | Intron | NP_001295081.1 | ||||
NM_001308153.1 | Intron | NP_001295082.1 | ||||
NM_014629.3 | Intron | NP_055444.2 | ||||
XM_005266041.3 | Intron | XP_005266098.1 | ||||
XM_011534767.2 | Intron | XP_011533069.1 | ||||
XM_011534768.1 | Intron | XP_011533070.1 | ||||
XM_011534770.2 | Intron | XP_011533072.1 | ||||
XM_017014003.1 | Intron | XP_016869492.1 |