Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGCGACTCCTCAAGAGGCCGGTCCA[A/C]GTGGAAGTCATGGATGTGGGGGGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610076 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDK20 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CDK20 - cyclin dependent kinase 20 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039803.2 | 1283 | Missense Mutation | NP_001034892.1 | |||
NM_001170639.1 | 1283 | UTR 3 | NP_001164110.1 | |||
NM_001170640.1 | 1283 | UTR 3 | NP_001164111.1 | |||
NM_012119.4 | 1283 | Missense Mutation | NP_036251.2 | |||
NM_178432.3 | 1283 | Missense Mutation | NP_848519.1 | |||
XM_017014561.1 | 1283 | Missense Mutation | XP_016870050.1 | |||
XM_017014562.1 | 1283 | UTR 3 | XP_016870051.1 | |||
XM_017014563.1 | 1283 | Missense Mutation | XP_016870052.1 | |||
XM_017014564.1 | 1283 | UTR 3 | XP_016870053.1 |