Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTACAGTACTGACCAGGTTCTGCCT[A/G]GCAGGTTCCCAGGTCTAAAAGACAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
C9orf57 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
C9orf57 - chromosome 9 open reading frame 57 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001128618.1 | 241 | Nonsense Mutation | CAG,TAG | Q,* 101 | NP_001122090.1 | |
XM_011518225.2 | 241 | Nonsense Mutation | CAG,TAG | Q,* 51 | XP_011516527.1 |