Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTAATTAGTGCAGAAAATATCTAAT[A/G]CACTCAGTTTCTATATGATGGGAAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
MEDAG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MEDAG - mesenteric estrogen dependent adipogenesis | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TEX26 - testis expressed 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152325.1 | Intron | NP_689538.1 | ||||
XM_011534919.2 | Intron | XP_011533221.1 | ||||
XM_011534921.2 | Intron | XP_011533223.1 | ||||
XM_011534924.2 | Intron | XP_011533226.1 | ||||
XM_011534925.2 | Intron | XP_011533227.1 | ||||
XM_011534926.2 | Intron | XP_011533228.1 | ||||
XM_011534927.2 | Intron | XP_011533229.1 | ||||
XM_011534928.2 | Intron | XP_011533230.1 |
TEX26-AS1 - TEX26 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |