Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGCCGCGGGCGTGGGGCGCTTACC[G/T]GCGGCGTGGGCAGGCAGGCCCGCCG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 601632 | ||||||||||||||||||||
Literature Links: |
LOC107987188 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
LOC107987188 - uncharacterized LOC107987188 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
POU4F1 - POU class 4 homeobox 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006237.3 | 356 | Missense Mutation | CAG,CCG | Q,P 41 | NP_006228.3 |
RNF219-AS1 - RNF219 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |