Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCATAGTCAACAGTTTTAGCAAAAG[G/A]ACTTTAGAAAATGACAATTTTTTTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||
Literature Links: |
USP12 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
USP12 - ubiquitin specific peptidase 12 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_182488.3 | Intron | NP_872294.2 | ||||
XM_005266282.2 | Intron | XP_005266339.1 | ||||
XM_011534956.2 | Intron | XP_011533258.1 | ||||
XM_017020416.1 | Intron | XP_016875905.1 |