Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605276 MIM: 605475 MIM: 614059 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AATK PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AATK - apoptosis associated tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001080395.2 | 3499 | Missense Mutation | CCG,TCG | P,S 1192 | NP_001073864.2 | |
NM_004920.2 | 3499 | Missense Mutation | CCG,TCG | P,S 1089 | NP_004911.2 | |
XM_006722193.2 | 3499 | Missense Mutation | CCG,TCG | P,S 1144 | XP_006722256.1 | |
XM_006722194.2 | 3499 | Missense Mutation | CCG,TCG | P,S 1142 | XP_006722257.1 | |
XM_006722195.2 | 3499 | Missense Mutation | CCG,TCG | P,S 1025 | XP_006722258.1 | |
XM_006722196.2 | 3499 | Missense Mutation | CCG,TCG | P,S 1144 | XP_006722259.1 | |
XM_011525509.2 | 3499 | Missense Mutation | CCG,TCG | P,S 1151 | XP_011523811.1 | |
XM_011525510.2 | 3499 | Missense Mutation | CCG,TCG | P,S 1136 | XP_011523812.1 | |
XM_011525511.2 | 3499 | Missense Mutation | CCG,TCG | P,S 970 | XP_011523813.1 | |
XM_017025421.1 | 3499 | Missense Mutation | CCG,TCG | P,S 1089 | XP_016880910.1 | |
XM_017025422.1 | 3499 | Missense Mutation | CCG,TCG | P,S 970 | XP_016880911.1 |
BAIAP2 - BAI1 associated protein 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR3065 - microRNA 3065 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR338 - microRNA 338 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR657 - microRNA 657 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |