Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCACTCTTCCCTCTCCACTGCCAC[A/G]AAAAATTGCTTGATGCCTTCCAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608546 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
EIF4A3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
EIF4A3 - eukaryotic translation initiation factor 4A3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014740.3 | 880 | Silent Mutation | TTC,TTT | F,F 255 | NP_055555.1 | |
XM_011525522.1 | 880 | Silent Mutation | TTC,TTT | F,F 234 | XP_011523824.1 |