Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGGAGATGATAATTCGACTACTAC[G/T]ACAGCGGTGGCAGCTGCAGCTTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600518 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERPINB4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SERPINB4 - serpin family B member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002974.3 | 1194 | Silent Mutation | GTA,GTC | V,V 349 | NP_002965.1 | |
NM_175041.1 | 1194 | Silent Mutation | GTA,GTC | V,V 328 | NP_778206.1 | |
XM_011526138.1 | 1194 | Silent Mutation | GTA,GTC | V,V 349 | XP_011524440.1 |