Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCCAAGCCCCACCGCGAGACCCGG[C/T]GAAGGCCGAGGGCCGGGTCCCGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612086 MIM: 601697 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HMSD PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HMSD - histocompatibility minor serpin domain containing | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SERPINB8 - serpin family B member 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001031848.1 | Intron | NP_001027018.1 | ||||
NM_001276490.1 | Intron | NP_001263419.1 | ||||
NM_002640.3 | Intron | NP_002631.3 | ||||
NM_198833.1 | Intron | NP_942130.1 | ||||
XM_006722484.2 | Intron | XP_006722547.1 | ||||
XM_011526025.1 | Intron | XP_011524327.1 | ||||
XM_017025790.1 | Intron | XP_016881279.1 | ||||
XM_017025791.1 | Intron | XP_016881280.1 | ||||
XM_017025792.1 | Intron | XP_016881281.1 |