Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCTGTTTACAGAGTGTGGTAATTT[C/T]GTTCTTTAGATTTGCAGAATTTATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603356 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CD164 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CD164 - CD164 molecule | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001142401.1 | 717 | Missense Mutation | CAA,CGA | Q,R 179 | NP_001135873.1 | |
NM_001142402.1 | 717 | Missense Mutation | CAA,CGA | Q,R 173 | NP_001135874.1 | |
NM_001142403.1 | 717 | Intron | NP_001135875.1 | |||
NM_001142404.1 | 717 | Intron | NP_001135876.1 | |||
NM_006016.4 | 717 | Missense Mutation | CAA,CGA | Q,R 192 | NP_006007.2 |