Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACCCTATTAAAAAACAGAACAACAA[C/G]AAGAAAACCCCTTTACAAAAAAAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEP85L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEP85L - centrosomal protein 85 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001042475.2 | 8058 | UTR 3 | NP_001035940.1 | |||
NM_001178035.1 | 8058 | UTR 3 | NP_001171506.1 | |||
NM_206921.2 | 8058 | Intron | NP_996804.2 | |||
XM_005266970.1 | 8058 | UTR 3 | XP_005267027.1 | |||
XM_005266971.1 | 8058 | UTR 3 | XP_005267028.1 | |||
XM_005266972.4 | 8058 | UTR 3 | XP_005267029.1 | |||
XM_006715475.3 | 8058 | UTR 3 | XP_006715538.1 | |||
XM_011535809.2 | 8058 | UTR 3 | XP_011534111.1 | |||
XM_011535810.2 | 8058 | UTR 3 | XP_011534112.1 | |||
XM_011535811.1 | 8058 | UTR 3 | XP_011534113.1 | |||
XM_017010846.1 | 8058 | UTR 3 | XP_016866335.1 | |||
XM_017010847.1 | 8058 | UTR 3 | XP_016866336.1 |