Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AACATCACTCACGAAGTCATCCTTG[A/G]CCCCGAGCCGCTTTGTCCTGTCCTT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 602958 | ||||||||||||||||||||
Literature Links: |
SGK1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
SGK1 - serum/glucocorticoid regulated kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001143676.1 | 1159 | Missense Mutation | GCC,GTC | A,V 437 | NP_001137148.1 | |
NM_001143677.1 | 1159 | Missense Mutation | GCC,GTC | A,V 370 | NP_001137149.1 | |
NM_001143678.1 | 1159 | Missense Mutation | GCC,GTC | A,V 356 | NP_001137150.1 | |
NM_001291995.1 | 1159 | Missense Mutation | GCC,GTC | A,V 298 | NP_001278924.1 | |
NM_005627.3 | 1159 | Missense Mutation | GCC,GTC | A,V 342 | NP_005618.2 | |
XM_011536071.1 | 1159 | Missense Mutation | GCC,GTC | A,V 437 | XP_011534373.1 |