Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTCAGCAAACCATTGCCTGGCATCT[C/T]TCTGCTGTACAACACAGGCTACATG
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 123838 | ||||||||||||||||||||
Literature Links: |
CCNC PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CCNC - cyclin C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001013399.1 | 1078 | Missense Mutation | AAA,AGA | K,R 126 | NP_001013417.1 | |
NM_005190.3 | 1078 | Missense Mutation | AAA,AGA | K,R 211 | NP_005181.2 | |
XM_006715594.1 | 1078 | Missense Mutation | AAA,AGA | K,R 211 | XP_006715657.1 | |
XM_011536232.2 | 1078 | Intron | XP_011534534.1 | |||
XM_017011436.1 | 1078 | Intron | XP_016866925.1 |
TSTD3 - thiosulfate sulfurtransferase (rhodanese)-like domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001195131.1 | 1078 | Intron | NP_001182060.1 | |||
XM_017010141.1 | 1078 | Intron | XP_016865630.1 |