Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACAAAGTCAGATTCGTGACTTTTT[C/T]GCAGCCGGTGGTGTGCTAACTTGGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605680 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BAZ1A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BAZ1A - bromodomain adjacent to zinc finger domain 1A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013448.2 | 3348 | Silent Mutation | GCA,GCG | A,A 1551 | NP_038476.2 | |
NM_182648.1 | 3348 | Silent Mutation | GCA,GCG | A,A 1519 | NP_872589.1 | |
XM_011536374.1 | 3348 | Silent Mutation | GCA,GCG | A,A 1458 | XP_011534676.1 | |
XM_011536376.2 | 3348 | Silent Mutation | GCA,GCG | A,A 1074 | XP_011534678.2 | |
XM_017020942.1 | 3348 | Silent Mutation | GCA,GCG | A,A 1042 | XP_016876431.1 |