Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAGCAGCTGGCTGCTCCCTTAGGAT[C/T]CTTGACTTTGGGGAGCTCTGGCTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 612811 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LRFN5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LRFN5 - leucine rich repeat and fibronectin type III domain containing 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152447.3 | 549 | Intron | NP_689660.2 | |||
XM_005267369.3 | 549 | UTR 5 | XP_005267426.1 | |||
XM_017021048.1 | 549 | Intron | XP_016876537.1 | |||
XM_017021049.1 | 549 | UTR 5 | XP_016876538.1 | |||
XM_017021050.1 | 549 | UTR 5 | XP_016876539.1 |