Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGCCAGATGTTCAGCAGCAGCC[A/G]GAGTTGGACCCTATGTCACGAGAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607466 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAB2B PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RAB2B - RAB2B, member RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163380.1 | 667 | Silent Mutation | TCC,TCT | S,S 167 | NP_001156852.1 | |
NM_032846.3 | 667 | Silent Mutation | TCC,TCT | S,S 213 | NP_116235.2 | |
XM_017021710.1 | 667 | Silent Mutation | TCC,TCT | S,S 168 | XP_016877199.1 | |
XM_017021711.1 | 667 | Silent Mutation | TCC,TCT | S,S 148 | XP_016877200.1 | |
XM_017021712.1 | 667 | Silent Mutation | TCC,TCT | S,S 144 | XP_016877201.1 |